Assay Details
Gene Symbol: Ufm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126080.2 | NP_001119552.1 | 1 - 2 | 82 | 68 | |
GenBank mRNA | CB787557.1 | - | 1 - 2 | 120 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001126080.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
