Assay Details
Gene Symbol: Ufm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126080.2 | NP_001119552.1 | 2 - 3 | 142 | 104 | |
GenBank mRNA | BC091395.1 | - | 2 - 3 | 78 | 104 | |
CB787557.1 | - | 2 - 3 | 180 | 104 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC091395.1, NM_001126080.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
