Assay Details
Gene Symbol: Ufm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001126080.2 | NP_001119552.1 | 5 - 6 | 271 | 92 | |
GenBank mRNA | BC091395.1 | - | 5 - 6 | 207 | 92 | |
CB787557.1 | - | 5 - 6 | 309 | 92 | ||
CV112030.1 | - | 4 - 5 | 161 | 92 | ||
FQ229705.1 | - | 5 - 6 | 242 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
