Assay Details
Gene Symbol: Slc1a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108973.1 | NP_001102443.1 | 4 - 5 | 717 | 72 | |
XM_008763892.1 | XP_008762114.1 | 4 - 5 | 646 | 72 | ||
GenBank mRNA | JF422064.1 | - | 4 - 5 | 519 | 72 | |
JF422065.1 | - | 3 - 4 | 303 | 72 | ||
JF422066.1 | - | 4 - 5 | 519 | 72 | ||
JF422067.1 | - | 4 - 5 | 519 | 72 | ||
JF422068.1 | - | 4 - 5 | 519 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
