Assay Details
Gene Symbol: Slc1a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108973.1 | NP_001102443.1 | 8 - 9 | 1472 | 64 | |
GenBank mRNA | JF422064.1 | - | 8 - 9 | 1109 | 64 | |
JF422065.1 | - | 7 - 8 | 1058 | 64 | ||
JF422068.1 | - | 8 - 9 | 1274 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
