Assay Details
Gene Symbol: Artn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053397.1 | NP_445849.1 | 4 - 5 | 454 | 90 | |
XM_006238684.2 | XP_006238746.1 | 2 - 3 | 3968 | 90 | ||
XM_006238685.2 | XP_006238747.1 | 3 - 4 | 3749 | 90 | ||
XM_006238687.2 | XP_006238749.1 | 3 - 4 | 3704 | 90 | ||
GenBank mRNA | AY230412.1 | - | 2 - 3 | 197 | 90 | |
BC079078.1 | - | 4 - 5 | 454 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
