Assay Details
Gene Symbol: Acot11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271383.1 | NP_001258312.1 | 8 - 9 | 1005 | 68 | |
XM_008763851.1 | XP_008762073.1 | 8 - 9 | 1025 | 68 | ||
XM_008763852.1 | XP_008762074.1 | 8 - 9 | 966 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
