Assay Details
Gene Symbol: Eri3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238738.2 | XP_006238800.1 | 6 - 7 | 3112 | 97 | |
XM_006238740.2 | XP_006238802.1 | 6 - 7 | 477 | 97 | ||
XM_233435.6 | XP_233435.5 | 7 - 8 | 1061 | 97 | ||
GenBank mRNA | BC167080.1 | - | 6 - 7 | 701 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
