Assay Details
Gene Symbol: Eri3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238735.2 | XP_006238797.1 | 9 - 10 | 1302 | 86 | |
XM_006238736.2 | XP_006238798.1 | 9 - 10 | 1296 | 86 | ||
XM_006238737.2 | XP_006238799.1 | 10 - 11 | 1111 | 86 | ||
XM_006238738.2 | XP_006238800.1 | 7 - 8 | 3211 | 86 | ||
XM_006238740.2 | XP_006238802.1 | 7 - 8 | 576 | 86 | ||
XM_006238741.2 | XP_006238803.1 | 9 - 10 | 998 | 86 | ||
XM_008764085.1 | XP_008762307.1 | 9 - 10 | 3321 | 86 | ||
XM_008764086.1 | XP_008762308.1 | 8 - 9 | 1297 | 86 | ||
XM_233435.6 | XP_233435.5 | 8 - 9 | 1160 | 86 | ||
GenBank mRNA | BC167080.1 | - | 7 - 8 | 800 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
