Assay Details
Gene Symbol: Ccnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108110.1 | NP_001101580.1 | 2 - 3 | 248 | 122 | |
XM_008765745.1 | XP_008763967.1 | 2 - 3 | 408 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
