Assay Details
Gene Symbol: Racgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108112.1 | NP_001101582.1 | 4 - 5 | 610 | 112 | |
XM_006257400.2 | XP_006257462.1 | 4 - 5 | 557 | 112 | ||
GenBank mRNA | BC166473.1 | - | 4 - 5 | 510 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
