Assay Details
Gene Symbol: Pip5k1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042621.1 | NP_001036086.1 | 10 - 11 | 1555 | 68 | |
XM_008761315.1 | XP_008759537.1 | 10 - 11 | 1684 | 68 | ||
XM_008761316.1 | XP_008759538.1 | 10 - 11 | 1681 | 68 | ||
GenBank mRNA | AY724476.1 | - | 10 - 11 | 1555 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
