Assay Details
Gene Symbol: Slc22a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107707.1 | NP_001101177.1 | 12 - 13 | 1825 | 75 | |
XM_006233050.2 | XP_006233112.1 | 11 - 12 | 4219 | 75 | ||
GenBank mRNA | FQ211559.1 | - | 8 - 9 | 1406 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P140266 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P1059229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 293914 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355730 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804640 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 376821 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 689014 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231639 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 827601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 789172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 714459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 16149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 479961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859476 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 34254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 240629 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669118 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 965550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 947106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 48669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775018 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 920504 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 702483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 324716 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 629421 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 222267 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 112175 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688615 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 480195 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 353549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524246 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 687284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 186340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 436913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875109 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 48278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 46461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 78243 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 122165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61444 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P140266 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1059229 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P1059229 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P140266 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P140266 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P1059229 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P140266 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 83309 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 71152 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10825497 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825502 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825498 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825500 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825505 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825504 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825496 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825501 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825507 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825506 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825499 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825503 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10825495 | - | - |
Rat Genome 230 2.0 Array | 1398435_at | - | - |
Rat Genome 230 2.0 Array | 1398435_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398435_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1398435_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0200029879.rn.1 | TC0200004341.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200029881.rn.1 | TC0200004341.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0200029880.rn.1 | TC0200004341.rn.1 | RTA TC |
Rat Expression Set 230 | 1398435_PM_at | - | - |
Rat Expression Set 230 | 1398435_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10825495 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21874496 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359319 | - | - |
