Assay Details
Gene Symbol: Slc22a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107707.1 | NP_001101177.1 | 8 - 9 | 1356 | 67 | |
XM_006233050.2 | XP_006233112.1 | 7 - 8 | 3750 | 67 | ||
GenBank mRNA | FQ211559.1 | - | 4 - 5 | 937 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
