Assay Details
Gene Symbol: Sec22b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025686.1 | NP_001020857.1 | 2 - 3 | 288 | 85 | |
GenBank mRNA | BC098725.1 | - | 2 - 3 | 288 | 85 | |
FQ211503.1 | - | 313 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
