Assay Details
Gene Symbol: Mtmr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005761.1 | NP_001005761.1 | 1 - 2 | 430 | 143 | |
GenBank mRNA | AF526269.1 | - | 1 - 2 | 42 | 143 | |
BC083644.1 | - | 1 - 2 | 430 | 143 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
