Assay Details
Gene Symbol: Blk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025751.1 | NP_001020922.1 | 8 - 9 | 1102 | 69 | |
XM_006252188.2 | XP_006252250.1 | 9 - 10 | 1496 | 69 | ||
XM_008770769.1 | XP_008768991.1 | 8 - 9 | 1523 | 69 | ||
XM_008770770.1 | XP_008768992.1 | 7 - 8 | 762 | 69 | ||
GenBank mRNA | BC098683.1 | - | 8 - 9 | 1102 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
