Assay Details
Gene Symbol: Hibadh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022243.1 | NP_071579.1 | 3 - 4 | 392 | 153 | |
GenBank mRNA | BC127442.1 | - | 3 - 4 | 378 | 153 | |
J04628.1 | - | 3 - 4 | 393 | 153 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
