Assay Details
Gene Symbol: Atp6v1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108318.2 | NP_001101788.1 | 10 - 11 | 1331 | 83 | |
XM_006248348.2 | XP_006248410.1 | 10 - 11 | 1532 | 83 | ||
XM_006248349.2 | XP_006248411.1 | 10 - 11 | 1523 | 83 | ||
GenBank mRNA | FQ211993.1 | - | 10 - 11 | 1364 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
