Assay Details
Gene Symbol: Ildr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127536.3 | NP_001121008.2 | 1 - 2 | 142 | 72 | |
XM_006248394.2 | XP_006248456.1 | 1 - 2 | 211 | 72 | ||
GenBank mRNA | BC166709.1 | - | 1 - 2 | 142 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
