Assay Details
Gene Symbol: Bace2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002802.1 | NP_001002802.1 | 2 - 3 | 392 | 74 | |
XM_006248153.2 | XP_006248215.1 | 2 - 3 | 446 | 74 | ||
GenBank mRNA | BC166597.1 | - | 2 - 3 | 406 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
