Assay Details
Gene Symbol: Bace2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002802.1 | NP_001002802.1 | 5 - 6 | 873 | 63 | |
XM_006248153.2 | XP_006248215.1 | 5 - 6 | 927 | 63 | ||
GenBank mRNA | BC166597.1 | - | 5 - 6 | 887 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
