Assay Details
Gene Symbol: Sec22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768692.1 | XP_008766914.1 | 1 - 2 | 467 | 104 | |
XM_008768693.1 | XP_008766915.1 | 1 - 2 | 469 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
