Assay Details
Gene Symbol: Sec22a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_057147.2 | NP_476488.2 | 6 - 7 | 756 | 101 | |
XM_006248419.2 | XP_006248481.1 | 7 - 8 | 958 | 101 | ||
XM_006248420.2 | XP_006248482.1 | 6 - 7 | 993 | 101 | ||
XM_008768692.1 | XP_008766914.1 | 6 - 7 | 1205 | 101 | ||
GenBank mRNA | BC081746.1 | - | 6 - 7 | 756 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
