Assay Details
Gene Symbol: Abhd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123352.1 | NP_001116824.1 | 4 - 5 | 551 | 141 | |
GenBank mRNA | BC088235.1 | - | 4 - 5 | 544 | 141 | |
BC161919.1 | - | 4 - 5 | 551 | 141 |
Gene Symbol: LOC103693563 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008768673.1 | XP_008766895.1 | 4 - 5 | 681 | 141 | |
XM_008768674.1 | XP_008766896.1 | 4 - 5 | 1288 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
