Assay Details
Gene Symbol: Arhgap36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257554.2 | XP_006257616.2 | 5 - 6 | 906 | 81 | |
XM_006257555.2 | XP_006257617.1 | 5 - 6 | 929 | 81 | ||
XM_229133.8 | XP_229133.5 | 5 - 6 | 906 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
