Assay Details
Gene Symbol: Zfp280c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006257541.2 | XP_006257603.1 | 2427 | 67 | ||
XM_006257542.2 | XP_006257604.1 | 2326 | 67 | |||
XM_008773539.1 | XP_008771761.1 | 19 - 20 | 2314 | 67 | ||
XM_008773540.1 | XP_008771762.1 | 2283 | 67 | |||
XM_008773541.1 | XP_008771763.1 | 18 - 19 | 2278 | 67 | ||
XM_008773542.1 | XP_008771764.1 | 17 - 18 | 3308 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
