Assay Details
Gene Symbol: Itgb3bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013213.1 | NP_001013231.1 | 1 - 2 | 335 | 71 | |
GenBank mRNA | BC086363.1 | - | 1 - 2 | 335 | 71 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC086363.1, NM_001013213.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
