Assay Details
Gene Symbol: Ror1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108671.1 | NP_001102141.1 | 2 - 3 | 582 | 91 | |
XM_008763887.1 | XP_008762109.1 | 1 - 2 | 1432 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
