Assay Details
Gene Symbol: Ror1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108671.1 | NP_001102141.1 | 7 - 8 | 1601 | 86 | |
XM_008763887.1 | XP_008762109.1 | 6 - 7 | 2451 | 86 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
