Assay Details
Gene Symbol: Rinl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106241.1 | NP_001099711.1 | 2 - 3 | 155 | 64 | |
XM_006228653.2 | XP_006228715.1 | 2 - 3 | 407 | 64 | ||
XM_006228654.2 | XP_006228716.1 | 2 - 3 | 395 | 64 | ||
XM_006228655.2 | XP_006228717.1 | 2 - 3 | 407 | 64 | ||
XM_006228656.2 | XP_006228718.1 | 2 - 3 | 407 | 64 | ||
XM_008759121.1 | XP_008757343.1 | 2 - 3 | 240 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
