Assay Details
Gene Symbol: Rinl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106241.1 | NP_001099711.1 | 8 - 9 | 1189 | 64 | |
XM_006228653.2 | XP_006228715.1 | 8 - 9 | 1441 | 64 | ||
XM_006228654.2 | XP_006228716.1 | 8 - 9 | 1429 | 64 | ||
XM_006228655.2 | XP_006228717.1 | 8 - 9 | 1441 | 64 | ||
XM_006228656.2 | XP_006228718.1 | 8 - 9 | 1441 | 64 | ||
XM_006228657.2 | XP_006228719.1 | 7 - 8 | 1164 | 64 | ||
XM_008759120.1 | XP_008757342.1 | 7 - 8 | 1306 | 64 | ||
XM_008759121.1 | XP_008757343.1 | 8 - 9 | 1274 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
