Assay Details
Gene Symbol: Pak4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106238.1 | NP_001099708.1 | 1 - 2 | 111 | 72 | |
GenBank mRNA | BC168940.1 | - | 1 - 2 | 121 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC168940.1, NM_001106238.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
