Assay Details
Gene Symbol: Dcaf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107935.1 | NP_001101405.1 | 4 - 5 | 1092 | 92 | |
XM_006238066.2 | XP_006238128.1 | 3 - 4 | 930 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
MicroArray Probes:
