Assay Details
Gene Symbol: B4galt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053287.1 | NP_445739.1 | 5 - 6 | 1289 | 79 | |
GenBank mRNA | CB741712.1 | - | 1 - 2 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
epithelial cell development
acute inflammatory response
lactose biosynthetic process
galactose metabolic process
protein glycosylation
protein N-linked glycosylation
cell adhesion
binding of sperm to zona pellucida
penetration of zona pellucida
negative regulation of cell population proliferation
glycoprotein biosynthetic process
oligosaccharide biosynthetic process
response to wounding
extracellular matrix organization
mammary gland development
wound healing
regulation of cell population proliferation
positive regulation of apoptotic process
development of secondary sexual characteristics
branching morphogenesis of an epithelial tube
leukocyte migration
regulation of cellular component movement
regulation of acrosome reaction
positive regulation of epithelial cell proliferation involved in wound healing
angiogenesis involved in wound healing
positive regulation of apoptotic process involved in mammary gland involution
molecular_function
beta-N-acetylglucosaminylglycopeptide beta-1,4-galactosyltransferase activity
N-acetyllactosamine synthase activity
lactose synthase activity
cytoskeletal protein binding
galactosyltransferase activity
protein kinase binding
manganese ion binding
UDP-galactosyltransferase activity
protein homodimerization activity
alpha-tubulin binding
metal ion binding
beta-tubulin binding
epithelial cell development
acute inflammatory response
lactose biosynthetic process
galactose metabolic process
protein glycosylation
protein N-linked glycosylation
cell adhesion
binding of sperm to zona pellucida
penetration of zona pellucida
negative regulation of cell population proliferation
glycoprotein biosynthetic process
oligosaccharide biosynthetic process
response to wounding
extracellular matrix organization
mammary gland development
wound healing
regulation of cell population proliferation
positive regulation of apoptotic process
development of secondary sexual characteristics
branching morphogenesis of an epithelial tube
leukocyte migration
regulation of cellular component movement
regulation of acrosome reaction
positive regulation of epithelial cell proliferation involved in wound healing
angiogenesis involved in wound healing
positive regulation of apoptotic process involved in mammary gland involution
molecular_function
beta-N-acetylglucosaminylglycopeptide beta-1,4-galactosyltransferase activity
N-acetyllactosamine synthase activity
lactose synthase activity
cytoskeletal protein binding
galactosyltransferase activity
protein kinase binding
manganese ion binding
UDP-galactosyltransferase activity
protein homodimerization activity
alpha-tubulin binding
metal ion binding
beta-tubulin binding
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P879764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257102 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 431172 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 866956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255767 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 759438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891157 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 855499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458116 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 514659 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 573237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 177216 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 80337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 726019 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 257585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103167 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 674398 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 716370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 765274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 152763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 4169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 652764 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 354207 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 904926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 811866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 361938 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7851 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 102442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43686 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 126505 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 119548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150296 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P879764 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P879764 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P879764 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P773945 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P879764 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 37583 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 45822 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 10887 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 29042 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10876051 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876048 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876049 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876047 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876046 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10876050 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10876045 | - | - |
Rat Genome 230 2.0 Array | 1375250_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375250_at | - | - |
Rat Genome 230 2.0 Array | 1375755_at | - | - |
Rat Genome 230 2.0 Array | 1371073_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375755_PM_at | - | - |
Rat Genome 230 2.0 Array | 1371073_at | - | - |
Affymetrix HT Rat Focus Array | 1375250_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1371073_at | - | - |
Affymetrix HT Rat Focus Array | 1375250_at | - | - |
Affymetrix HT Rat Focus Array | 1375755_at | - | - |
Affymetrix HT Rat Focus Array | 1371073_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1375755_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500018772.rn.1 | TC0500002556.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018774.rn.1 | TC0500002556.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500018771.rn.1 | TC0500002556.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10876045 | - | - |
Rat Genome U34 Set | rc_AA818683_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22364854 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1372751 | - | - |
