Assay Details
Gene Symbol: Galt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013089.2 | NP_001013107.1 | 7 - 8 | 763 | 101 | |
XM_006238033.2 | XP_006238095.1 | 7 - 8 | 687 | 101 | ||
XM_008763615.1 | XP_008761837.1 | 5 - 6 | 651 | 101 | ||
GenBank mRNA | BC098756.1 | - | 7 - 8 | 763 | 101 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
