Assay Details
Gene Symbol: Suv39h2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108883.1 | NP_001102353.1 | 3 - 4 | 1086 | 129 | |
XM_008771877.1 | XP_008770099.1 | 3 - 4 | 1216 | 129 | ||
XM_008771878.1 | XP_008770100.1 | 3 - 4 | 2290 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
