Assay Details
Gene Symbol: Camk1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107365.1 | NP_001100835.1 | 2 - 3 | 371 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
