Assay Details
Gene Symbol: Camk1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107365.1 | NP_001100835.1 | 9 - 10 | 982 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
