Assay Details
Gene Symbol: Trdmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001031643.1 | NP_001026813.1 | 8 - 9 | 923 | 80 | |
GenBank mRNA | BC098700.1 | - | 8 - 9 | 923 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
