Assay Details
Gene Symbol: Aasdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006250858.2 | XP_006250920.1 | 15 - 16 | 3429 | 65 | |
XM_008770108.1 | XP_008768330.1 | 11 - 12 | 2589 | 65 | ||
XM_008770112.1 | XP_008768334.1 | 7 - 8 | 2819 | 65 | ||
XM_008770113.1 | XP_008768335.1 | 8 - 9 | 1778 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
