Assay Details
Gene Symbol: Ube2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106006.1 | NP_001099476.1 | 5 - 6 | 614 | 65 | |
XM_006250934.2 | XP_006250996.1 | 5 - 6 | 811 | 65 | ||
XM_008770140.1 | XP_008768362.1 | 3 - 4 | 411 | 65 | ||
GenBank mRNA | FQ226880.1 | - | 5 - 6 | 606 | 65 | |
FQ233155.1 | - | 5 - 6 | 562 | 65 | ||
FQ234076.1 | - | 5 - 6 | 592 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
