Assay Details
Gene Symbol: Tnfaip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003748656.3 | XP_003748704.2 | 6 - 7 | 1998 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
