Assay Details
Gene Symbol: Il22ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003404.1 | NP_001003404.1 | 1 - 2 | 85 | 81 | |
XM_006227664.2 | XP_006227726.1 | 2 - 3 | 138 | 81 | ||
GenBank mRNA | AJ555485.1 | - | 1 - 2 | 107 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
