Assay Details
Gene Symbol: Il22ra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003404.1 | NP_001003404.1 | 3 - 4 | 402 | 62 | |
XM_006227664.2 | XP_006227726.1 | 4 - 5 | 455 | 62 | ||
GenBank mRNA | AJ555485.1 | - | 3 - 4 | 424 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
