Assay Details
Gene Symbol: Ust | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108458.1 | NP_001101928.1 | 7 - 8 | 1314 | 60 | |
XM_008758621.1 | XP_008756843.1 | 5 - 6 | 959 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P147549 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 508508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 154743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 41444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 214444 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446829 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871028 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 261022 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 326391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 696040 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 804308 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 723115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 271449 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 452055 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 10917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481265 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 344844 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 544875 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 867003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 669927 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 307933 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486416 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12507 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 487375 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 325535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 355062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 490304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 610191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123820 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 889388 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 682095 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739000 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 921794 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 306498 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 729979 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 581337 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 264559 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327100 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539735 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 735982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 42578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 549275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533774 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 967237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 406036 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486003 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 19567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 693789 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 351763 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365183 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 163677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 491373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868572 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 673459 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940936 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 169092 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245236 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 914566 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727510 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250590 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220628 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 813051 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 464360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 493133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 630738 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 275552 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 919691 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 819322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 70389 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806727 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 895627 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 775455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 26953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 403414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 364280 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 347431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40959 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 925845 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506038 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741377 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 763675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 594711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 424806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 102384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 817562 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 742770 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 398463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 235149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 323980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 603192 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562857 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 455587 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 691915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 849178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 672543 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 924663 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 846496 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 934448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739257 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913611 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 233455 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483058 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103286 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 582904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 40726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 969420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343376 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123368 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 365561 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 894642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435340 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 378237 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68515 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649742 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 65184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 576942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 308539 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656466 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 256364 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 421846 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170816 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814381 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613747 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 843666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595737 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 416406 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 871720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 880323 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520752 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205811 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 423091 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 104721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 882391 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 783958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 656441 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 397502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 913435 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 923112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 494365 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 732928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189860 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 422598 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 733934 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 911750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468645 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 67913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 628090 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 821987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 754946 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12594 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 662509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945949 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 659540 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3616 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 312115 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201969 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 793494 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 758990 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 940080 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 418264 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 90189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 859079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 933053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 868198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 232279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 410349 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 660057 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 958608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 599170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 259506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 101522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 159089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13348 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10112 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180128 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 139760 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 69462 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 96700 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 40858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 58988 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50761 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20777 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89319 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52906 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 76043 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 149254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 70366 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 19631 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 84194 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 137903 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 75423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 150453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 173894 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27812 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 111089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 42113 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P147549 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P147549 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P147549 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 28176 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 62940 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 11742 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 41772 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57214 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 1946 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84528 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 8388 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91410 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 46330 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10716672 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716671 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716673 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716674 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716669 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716675 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716670 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10716668 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10716667 | - | - |
Affymetrix Rat Clariom S Assay | PSR0100032321.rn.1 | TC0100003961.rn.1 | RTA TC |
Affymetrix Rat Gene 1.0 ST Array | 10716667 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21427750 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1367041 | - | - |
