Assay Details
Gene Symbol: Dlat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031025.1 | NP_112287.1 | 12 - 13 | 1640 | 78 | |
GenBank mRNA | BC107440.1 | - | 12 - 13 | 1640 | 78 | |
D00092.1 | - | 9 - 10 | 1144 | 78 | ||
D10655.1 | - | 11 - 12 | 1357 | 78 | ||
M16075.1 | - | 9 - 10 | 1144 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
