Assay Details
Gene Symbol: Cbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242965.2 | XP_006243027.1 | 12 - 13 | 2141 | 63 | |
XM_576396.6 | XP_576396.3 | 13 - 14 | 2273 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
