Assay Details
Gene Symbol: Bco2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001127712.1 | NP_001121184.1 | 2 - 3 | 260 | 69 | |
GenBank mRNA | AY325176.1 | - | 425 | 69 | ||
BC133725.1 | - | 2 - 3 | 260 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
