Assay Details
Gene Symbol: Tbcel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242859.2 | XP_006242921.1 | 1 - 2 | 2850 | 64 | |
XM_008766134.1 | XP_008764356.1 | 2 - 3 | 2813 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
